Learn More
Medchemexpress LLC Oncrasin-1 | 75629-57-1 | 98.2% | 269.73 | 100 MG
SDP
Catalog No. 5000392481
Encompass_Preferred
Oncrasin-1 is an RNA polymerase inhibitor that suppresses the phosphorylation of RNA polymerase II and the expression of intronless reporter genes in sensitive cells. It effectively kills various human lung cancer cells with K-Ras mutations. It leads to coaggregation of PKCι and splicing factors into megaspliceosomes, inducing malfunction in the RNA processing machinery. This anti-cancer agent can be studied in research for lung cancer.
  • RNA polymerase inhibitor
  • Suppresses phosphorylation of RNA polymerase II and expression of intronless reporter genes
  • Effectively kills human lung cancer cells with K-Ras mutations
  • Induces coaggregation of PKCι and splicing factors into megaspliceosomes
  • Causes malfunction in the RNA processing machinery
  • Anti-cancer agent for lung cancer research
  • Appears as a light yellow to khaki solid
Catalog No. 50-003-92481 Supplier Medchemexpress LLC Supplier No. HY16662100MG
Oncrasin-1 is an RNA polymerase inhibitor that suppresses the phosphorylation of RNA polymerase II and the expression of intronless reporter genes in sensitive cells. It effectively kills various human lung cancer cells with K-Ras mutations. It leads to coaggregation of PKCι and splicing factors into megaspliceosomes, inducing malfunction in the RNA processing machinery. This anti-cancer agent can be studied in research for lung cancer.
  • RNA polymerase inhibitor
  • Suppresses phosphorylation of RNA polymerase II and expression of intronless reporter genes
  • Effectively kills human lung cancer cells with K-Ras mutations
  • Induces coaggregation of PKCι and splicing factors into megaspliceosomes
  • Causes malfunction in the RNA processing machinery
  • Anti-cancer agent for lung cancer research
  • Appears as a light yellow to khaki solid