Learn More
Selleck Chemical LLC Ondansetron Hydrochloride Dihydrate S4748-50mg
SDP
Catalog No. 503109891
Encompass_Preferred
Ondansetron is a highly specific and selective serotonin 5-HT3 receptor antagonist (Ki 6 16 nM) with low affinity for dopamine receptors
Catalog No. 50-310-9891 Supplier Selleck Chemical LLC Supplier No. S474850MG
Ondansetron is a highly specific and selective serotonin 5-HT3 receptor antagonist (Ki 6 16 nM) with low affinity for dopamine receptors