Learn More
Selleck Chemical LLC Ondansetron HCl 10mg 99614-01-4 GR 38032F
SDP
Catalog No. 501361204
Encompass_Preferred
Ondansetron HCl (GR 38032F) is a serotonin 5-HT3 receptor antagonist, used to prevent nausea and vomiting caused by cancer chemotherapy, and radiation therapy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1204 Supplier Selleck Chemical LLC Supplier No. S139010MG
Ondansetron HCl (GR 38032F) is a serotonin 5-HT3 receptor antagonist, used to prevent nausea and vomiting caused by cancer chemotherapy, and radiation therapy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.