Learn More
Apexbio Technology LLC Ondansetron hydrochloride dihydrate 103639-04-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B12045.1
Encompass_Preferred
Ondansetron hydrochloride dihydrate (CAS 103639-04-9) is a small molecule antagonist targeting the serotonin (5-HT3) receptor a ligand-gated ion channel prominent in the central and peripheral nervous systems Through blockade of 5-HT3 receptor signaling ondansetron inhibits serotonin-induced emetic responses Due to this mechanism it is widely utilized in biomedical research investigating serotonin-related signaling pathways and nausea mechanisms particularly in contexts involving chemotherapy-induced emesis
Catalog No. 50-000-63233