Learn More
Selleck Chemical LLC Ondansetron HCl 50mg 99614-01-4 GR 38032F
SDP
Supplier:  Selleck Chemical LLC S139050MG
Encompass_Preferred
Ondansetron HCl (GR 38032F) is a serotonin 5-HT3 receptor antagonist, used to prevent nausea and vomiting caused by cancer chemotherapy, and radiation therapy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-629-2