Learn More
Medchemexpress LLC Ondansetron (hydrochloride) | 99614-01-4 | 99.4% | C18H20ClN3O | 1 G
SDP
Catalog No. 5000432933
Encompass_Preferred
Ondansetron hydrochloride is a highly selective 5-HT3 receptor antagonist. It exerts its antiemetic effect by antagonizing 5-HT receptors located in localized neurons in the peripheral and central nervous systems, inhibiting nausea and vomiting induced by chemotherapy and radiotherapy.
  • Highly selective 5-HT3 receptor antagonist
  • Antagonizes 5-HT receptors to exert antiemetic effects
  • Inhibits nausea and vomiting induced by chemotherapy and radiotherapy
  • Can exhibit anti-inflammatory effects through the 5-HT3 receptor
Catalog No. 50-004-32933 Supplier Medchemexpress LLC Supplier No. HYB00021G
Ondansetron hydrochloride is a highly selective 5-HT3 receptor antagonist. It exerts its antiemetic effect by antagonizing 5-HT receptors located in localized neurons in the peripheral and central nervous systems, inhibiting nausea and vomiting induced by chemotherapy and radiotherapy.
  • Highly selective 5-HT3 receptor antagonist
  • Antagonizes 5-HT receptors to exert antiemetic effects
  • Inhibits nausea and vomiting induced by chemotherapy and radiotherapy
  • Can exhibit anti-inflammatory effects through the 5-HT3 receptor