Learn More
Medchemexpress LLC Ondansetron hydrochloride | 99614-01-4 | MFCD00764297 | 99.4% | C18H20ClN3O | 100 MG
SDP
Catalog No. 5000432227
Encompass_Preferred
Ondansetron hydrochloride (GR 38032; SN 307) is a highly selective 5-HT3 receptor antagonist. It exerts its antiemetic effect by antagonizing 5-HT receptors in the peripheral and central nervous systems, inhibiting nausea and vomiting induced by chemotherapy and radiotherapy.
  • Highly selective 5-HT3 receptor antagonist (IC50: 103 pM)
  • Exerts antiemetic effect
  • Inhibits nausea and vomiting from chemotherapy and radiotherapy
  • Reduces EAAT3 response to L-glutamate (IC50: 8.57 μM)
  • Exhibits anti-inflammatory effects through 5-HT3 receptor
Catalog No. 50-004-32227 Supplier Medchemexpress LLC Supplier No. HYB0002100MG
Ondansetron hydrochloride (GR 38032; SN 307) is a highly selective 5-HT3 receptor antagonist. It exerts its antiemetic effect by antagonizing 5-HT receptors in the peripheral and central nervous systems, inhibiting nausea and vomiting induced by chemotherapy and radiotherapy.
  • Highly selective 5-HT3 receptor antagonist (IC50: 103 pM)
  • Exerts antiemetic effect
  • Inhibits nausea and vomiting from chemotherapy and radiotherapy
  • Reduces EAAT3 response to L-glutamate (IC50: 8.57 μM)
  • Exhibits anti-inflammatory effects through 5-HT3 receptor