Learn More
Medchemexpress LLC Ondansetron hydrochloride dihydrate | 103639-04-9 | 99.9% | C18H24ClN3O3 | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0002A5G
Encompass_Preferred
Ondansetron hydrochloride dihydrate is a highly selective 5-HT3 receptor antagonist with an IC50 value of 103 pM. This compound exerts its antiemetic effect by antagonizing 5-HT3 receptors located in localized neurons in the peripheral and central nervous systems, which allows it to inhibit nausea and vomiting induced by chemotherapy and radiotherapy.
  • Highly selective 5-HT3 receptor antagonist
  • Antagonizes 5-HT3 receptors in peripheral and central nervous systems
  • Inhibits nausea and vomiting induced by chemotherapy and radiotherapy
Catalog No. 50-004-32934