Learn More
Medchemexpress LLC Ondansetron hydrochloride (GR 38032 hydrochloride) | 99614-01-4 | 99.4% | 50 MG
SDP
Catalog No. 5000432513
Encompass_Preferred
Ondansetron hydrochloride is a highly selective 5-HT3 receptor antagonist that inhibits nausea and vomiting induced by chemotherapy and radiotherapy. It acts by antagonizing 5-HT receptors in the peripheral and central nervous systems. This product is for research use only.
  • Highly selective 5-HT3 receptor antagonist
  • Inhibits nausea and vomiting caused by chemotherapy and radiotherapy
  • Exerts antiemetic effect through 5-HT receptor antagonism
  • Reduces EAAT3 response to L-glutamate in vitro
  • Exhibits anti-inflammatory effects in vivo
  • Purity of 99.4%
Catalog No. 50-004-32513 Supplier Medchemexpress LLC Supplier No. HYB000250MG
Ondansetron hydrochloride is a highly selective 5-HT3 receptor antagonist that inhibits nausea and vomiting induced by chemotherapy and radiotherapy. It acts by antagonizing 5-HT receptors in the peripheral and central nervous systems. This product is for research use only.
  • Highly selective 5-HT3 receptor antagonist
  • Inhibits nausea and vomiting caused by chemotherapy and radiotherapy
  • Exerts antiemetic effect through 5-HT receptor antagonism
  • Reduces EAAT3 response to L-glutamate in vitro
  • Exhibits anti-inflammatory effects in vivo
  • Purity of 99.4%