Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ondansetron Related Compound D, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023016 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-3016 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-3016 Supplier MilliporeSigma Supelco Supplier No. PHR2221100MG
Add to Cart
Add to Cart

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

TRUSTED_SUSTAINABILITY

Specifications

CAS 99614-64-9
Molecular Formula C14H13NO
Linear Formula C14H13NO
MDL Number MFCD08063682
Synonym 9-Methyl-3-methylene-1,2,3,9-tetrahydro-4 H-carbazol-4-one; 1,2,3,9-Tetrahydro-9-methyl-3-methylene-4 H-carbazol-4-one
Formula Weight 211.26
Grade Certified Reference Material
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.