Learn More
Cayman Chemical Ondansetron-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2699607-85-5, 1 mg
SDP
Catalog No. 502933023
Encompass_Preferred
An internal standard for the quantification of ondansetron by GC- or LC-MS
Catalog No. 50-293-3023 Supplier Cayman Chemical Supplier No. 257541 MG
An internal standard for the quantification of ondansetron by GC- or LC-MS