Learn More
Cayman Chemical Ondnsetron-13C-d3 500ug
SDP
Catalog No. 502936516
Encompass_Preferred
An internal standard for the quantification of ondansetron by GC- or LC-MS
Catalog No. 50-293-6516 Supplier Cayman Chemical Supplier No. 25754500 UG
An internal standard for the quantification of ondansetron by GC- or LC-MS