Learn More
World Precision Instrument One-Channel Free Radical Analyzer
SDP
Catalog No. 502521046
Encompass
Fast, reliable, real-time detection – measure redox-reactive species
Real-time detection and measurement of a variety of redox-reactive species is fast and easy using the electrochemical (amperometric) detection principle employed in the  TBR4100. This optically isolated four-channel free radical analyzer has ultra low noise and independently operated channels.
Catalog No. 50-252-1046 Supplier World Precision Instrument Supplier No. TBR1025
Add to Cart
Add to Cart
Request a Quote
Fast, reliable, real-time detection – measure redox-reactive species
Real-time detection and measurement of a variety of redox-reactive species is fast and easy using the electrochemical (amperometric) detection principle employed in the  TBR4100. This optically isolated four-channel free radical analyzer has ultra low noise and independently operated channels.