Learn More
Braintree Scientific One Directional Manifold Assembly for (3) anesthetic devices
SDP
Catalog No. 501955014
Encompass
One Directional Manifold AssemblyThe one directional manifold works like the traditional two directional manifold but allows placement versatility. The one directional manifold can be used to deliver fresh anesthetic gases or collect waste anesthetic gases (WAG). Gray standard, white recommended for WAG. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5014 Supplier Braintree Scientific Supplier No. M13
 Restricted Item
Add to Cart
Add to Cart
One Directional Manifold AssemblyThe one directional manifold works like the traditional two directional manifold but allows placement versatility. The one directional manifold can be used to deliver fresh anesthetic gases or collect waste anesthetic gases (WAG). Gray standard, white recommended for WAG. **Sold for research purposes only and not for human use**