Learn More
Grainger One Hole Paper Punch, 6 Sheet
Catalog No. NC2735825
Encompass_Preferred
The Grainger 2WFT9 One Hole Paper Punch is a manual paper punch designed for punching holes in up to 6 sheets of paper at a time. It features an all-steel construction with black finish and rubber base to prevent slipping. The punching mechanism allows for punching of 6 sheets of 20lb paper and has an adjustable paper guide and die-cast handle. This single hole punch creates holes sized for use with standard brads and has a hole diameter of 5/16 inches. It is lightweight, weighing 0.75 lbs. Overall dimensions are 3-1/2 inches length by 2-3/4 inches width by 4-3/4 inches height.
Catalog No. NC2735825 Supplier Grainger Supplier No. 2WFT9
The Grainger 2WFT9 One Hole Paper Punch is a manual paper punch designed for punching holes in up to 6 sheets of paper at a time. It features an all-steel construction with black finish and rubber base to prevent slipping. The punching mechanism allows for punching of 6 sheets of 20lb paper and has an adjustable paper guide and die-cast handle. This single hole punch creates holes sized for use with standard brads and has a hole diameter of 5/16 inches. It is lightweight, weighing 0.75 lbs. Overall dimensions are 3-1/2 inches length by 2-3/4 inches width by 4-3/4 inches height.