Learn More
Bachem Pentagastrin
SDP
Supplier:  Bachem 4002096.0100
Encompass_Preferred
Pentagastrin, Boc-b-Ala-Trp-Met-Asp-Phe-NH2, 5534-95-2, C37H49N7O9S, Mr 767.90, 100mg. ICI 50123, AY 6608, NSC 367746, Boc-(b-Ala13)-Gastrin (13-17), Boc-b-Ala-CCK-4. One of the most potent pentapeptides having the same qualitative effects as the parent hormone gastrin I. The effect of this cholecystokinin agonist has been studied in patients with generalized anxiety disorder. Pentagastrin is used as a diagnostic for the evaluation of gastric acid secretory function (pentagastrin test).
Catalog No. 50-194-6921