Learn More
Sigma Aldrich Fine Chemicals Biosciences One piece dry box gloves a
Catalog No. 502905314
Encompass
Butyl for highest impermeability to water vapor gases and toxic fluidsHypalon(R) for resistance to abrasion oxidizing chemicals ozone and UVIdeally suited for the handling of ketones esters acetone benzene ethanol and tolueneStays flexible at extreme temperaturesCan withstand VHPHas the highest impermeability to water vapor and is impermeable to most gasesHolds up well to physical stresses such as oxidation solvents and chemicals
Catalog No. 50-290-5314 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1873211PAK
Butyl for highest impermeability to water vapor gases and toxic fluidsHypalon(R) for resistance to abrasion oxidizing chemicals ozone and UVIdeally suited for the handling of ketones esters acetone benzene ethanol and tolueneStays flexible at extreme temperaturesCan withstand VHPHas the highest impermeability to water vapor and is impermeable to most gasesHolds up well to physical stresses such as oxidation solvents and chemicals