Learn More
Waters Corp Waters Fitting, One-Piece, 10-32, UPLC
Catalog No. 502039504 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This one-piece fitting is designed for use in UPLC systems. It is compatible with various column managers, column heaters, and UPLC instrumentation, offering reliable connections for high-performance liquid chromatography applications. Each package contains 10 units.
  • One-piece design for secure connections
  • Features 10-32 thread specification
  • Specifically for UPLC system types
  • Compatible with UPLC column managers and column heaters
  • Works with a range of UPLC instruments, including H-Class, I-Class, and 2D systems
  • Provided as a 10-unit package
  • Replacement as required to maintain system performance
Catalog No. 50-203-9504 Supplier Waters Corp Supplier No. 700004841
This one-piece fitting is designed for use in UPLC systems. It is compatible with various column managers, column heaters, and UPLC instrumentation, offering reliable connections for high-performance liquid chromatography applications. Each package contains 10 units.
  • One-piece design for secure connections
  • Features 10-32 thread specification
  • Specifically for UPLC system types
  • Compatible with UPLC column managers and column heaters
  • Works with a range of UPLC instruments, including H-Class, I-Class, and 2D systems
  • Provided as a 10-unit package
  • Replacement as required to maintain system performance