Learn More
LabScientific Heavy cardboard slide holders with thumb grooves for easy slide removal. 100/Pack
Catalog No. NC0354591
Encompass
Heavy cardboard holders with thumb grooves for easy slide removal. Each holds standart 3 x 1 in. (75 x 25mm) slides. Measures: One place holder measure: 95 x 4 x 5 mm. Two place measure: 13 x 79 x 5 mm. Four place measure: 15 x 12 x 5 mm. 100/Pack
Catalog No. NC0354591 Supplier LabScientific Supplier No. SM002
Heavy cardboard holders with thumb grooves for easy slide removal. Each holds standart 3 x 1 in. (75 x 25mm) slides. Measures: One place holder measure: 95 x 4 x 5 mm. Two place measure: 13 x 79 x 5 mm. Four place measure: 15 x 12 x 5 mm. 100/Pack