Learn More
Med Vet International One Rack Test Tube Rack, 72-Places, 16mm, Blue, J0352X, Each
Catalog No. 5000584772
Encompass_Preferred
Two desirable features have been combined to create the Smooth Rack. First, when using a Smooth Rack tubes will never slip into an adjacent position. Second, the Smooth Rack is versatile. It accommodates a wide range of tubes — 10 to 16mm in diameter.• 72 tube capacity• 9.7” x 5.0” x 2.6”• Autoclavable Polypropylene • Made in USA• Patent Pending
Catalog No. 50-005-84772 Supplier Med Vet International Supplier No. JOR352X
Two desirable features have been combined to create the Smooth Rack. First, when using a Smooth Rack tubes will never slip into an adjacent position. Second, the Smooth Rack is versatile. It accommodates a wide range of tubes — 10 to 16mm in diameter.• 72 tube capacity• 9.7” x 5.0” x 2.6”• Autoclavable Polypropylene • Made in USA• Patent Pending