Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS One Solution st LpH™ III Disinfectant System

Catalog No. 17463011 Shop All STERIS Corporation Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
17-463-011 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-463-011 Supplier STERIS Supplier No. 1S16CX
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Formulated for the disinfection of hard surfaces, One Solution combines a sterile unit dose pouch with a gallon of USP Water For Injection (WFI)

  • Liquid concentrates specifically formulated for the disinfection of hard surfaces, non-porous surfaces
  • Phenolic active ingredients provide broad spectrum antimicrobial efficacy against common bacteria, without harming the surface
  • Use-dilution rate of 1:256, formulated to maintain an acidic pH (2.6 to 3)
  • Can be applied manually or sprayed
  • Ideal for use in a contamination control rotational program
  • Gamma irradiated and double bagged for easy introduction into aseptic areas
  • Each lot documented by a Certificate of Analysis that includes irradiation and sterility results

Specifications

Product Type Phenolic Disinfectant
Form Concentrated Liquid
Sterility Sterile
Concentration 22%
For Use With (Application) Disinfecting Hard, Non-Porous Surfaces
Includes 1 oz. Pouches (4), 1 Gallon WFI (4)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.