Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS One Solution st Vesphene™ III Disinfectant System

Catalog No. 19415146 Shop All STERIS Corporation Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
19-415-146 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-415-146 Supplier STERIS Supplier No. 1S15CX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A phenolic, alkaline disinfectant that combines a sterile unit dose pouch with a gallon of USP Water For Injection (WFI)

  • Specifically formulated for the routine disinfection of hard, non-porous surfaces
  • Phenolic active ingredients provide broad spectrum antimicrobial efficacy against common bacteria and fungi
  • Use-dilution of rate of 0.8% v/v (1:128), can be applied manually or sprayed
  • Can be used alone or in rotation with the LpH™ III Disinfectants
  • Includes an extensive documentation package to meet validation objectives
  • Supported by the STERIS Technical Services team for individualized application and validation assistance
  • Each lot documented by Certificate of Analysis

Specifications

Product Type Phenolic Disinfectant
Sterility Sterile
Composition Phenolic Active Ingredients
Concentration 0.8% v/v (1:128)
For Use With (Application) Routine Disinfection of Hard, Non-Porous Surfaces
Includes Unit Dose Pouch (1 oz.), USP Water For Injection (1 Gallon)
Quantity 4/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.