Learn More
STERIS One Solution st™ Water For Injection
Catalog No. 19415116 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
2 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-415-116 -
19-415-117 2 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-415-116 Supplier STERIS Supplier No. 1S0308WR
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
A diluent for surface sterilants, disinfectants, sanitizers, or cleaners used in cleanrooms or other critical environments. Meets USP specifications and is double bagged for easy entry into cleanrooms or aseptic areas.
  • Intended for diluting concentrated cleaners or disinfectants
  • Filtered, and filled in a controlled environment
  • Each package is double bagged and gamma irradiated for easy entry into controlled environments and aseptic core areas
  • Includes an extensive documentation package to meet validation objectives
  • Products meet United States Pharmacopeia (USP) Water For Injection (WFI) specifications
  • Supported by the STERIS Technical Services team for individualized application and validation assistance
Specifications
Product Type Water for Injection (WFI)
Sterility Sterile
Certifications/Compliance Meets United States Pharmacopeia (USP) Water For Injection (WFI) Specifications
Filtered Filtered
For Use With (Application) Diluting Concentrated Cleaners or Disinfectants
Packaging Double Bagged
Quantity 4 x 1 gallon