Learn More
MSC Rochester Midland Corporation One Step Disinfectant Cleaner, Concentrated, Lemon, 55 Gallon Drum
Catalog No. 502838071
Encompass
This concentrated one-step disinfectant cleaner is designed for effective cleaning, disinfecting, and deodorizing of hard, non-porous surfaces. It offers virucidal, bactericidal, fungicidal, and mildewstatic efficacy, making it suitable for a variety of applications.
  • One step disinfectant cleaner
  • Concentrated formula with hard water tolerance
  • Penetrates and cleans soil effectively
  • Disinfects and deodorizes surfaces
  • Can be applied with a mop and bucket, trigger sprayers, sponge, or by soaking
  • Effective against a wide range of bacteria, viruses (including HIV-1, Human Coronavirus, Influenza A, Hepatitis B, and Hepatitis C), and fungi like *Trichophyton mentagrophytes*
  • Controls *Aspergillus niger* mold and mildew odors
  • Reduces the hazard of cross-contamination on hard surfaces
Catalog No. 50-283-8071 Supplier MSC Supplier No. 41414640
This concentrated one-step disinfectant cleaner is designed for effective cleaning, disinfecting, and deodorizing of hard, non-porous surfaces. It offers virucidal, bactericidal, fungicidal, and mildewstatic efficacy, making it suitable for a variety of applications.
  • One step disinfectant cleaner
  • Concentrated formula with hard water tolerance
  • Penetrates and cleans soil effectively
  • Disinfects and deodorizes surfaces
  • Can be applied with a mop and bucket, trigger sprayers, sponge, or by soaking
  • Effective against a wide range of bacteria, viruses (including HIV-1, Human Coronavirus, Influenza A, Hepatitis B, and Hepatitis C), and fungi like *Trichophyton mentagrophytes*
  • Controls *Aspergillus niger* mold and mildew odors
  • Reduces the hazard of cross-contamination on hard surfaces