Learn More
Epigentek Inc EpiQuik One-Step DNA Hydrolysis Kit (96 samples)
SDP
Catalog No. NC0497553
Encompass_Preferred
The EpiQuik One-Step DNA Hydrolysis Kit is a complete set of essential components which enables the experimenter to rapidly hydrolyze DNA to deoxynucleosides with a process that can be performed in a single incubation.
Catalog No. NC0497553 Supplier Epigentek Inc Supplier No. P102396
The EpiQuik One-Step DNA Hydrolysis Kit is a complete set of essential components which enables the experimenter to rapidly hydrolyze DNA to deoxynucleosides with a process that can be performed in a single incubation.