Learn More
BPS Bioscience Inc ONE-Step Luciferase Assay System, 100ml
SDP
Catalog No. NC0835052
Encompass
The One-Step Luciferase Assay System is designed for high-throughput, sensitive quantitation of firefly luciferase activity in mammalian cell culture. The reagent consists of two components, a Luciferase Reagent Buffer (Component A) and Luciferase Reagent Substrate (Component B). Component A and Component B are combined to form a working solution that contains all the necessary reagents for cell lysis and luciferase quantitation.
Catalog No. NC0835052 Supplier BPS Bioscience Inc Supplier No. 606902
The One-Step Luciferase Assay System is designed for high-throughput, sensitive quantitation of firefly luciferase activity in mammalian cell culture. The reagent consists of two components, a Luciferase Reagent Buffer (Component A) and Luciferase Reagent Substrate (Component B). Component A and Component B are combined to form a working solution that contains all the necessary reagents for cell lysis and luciferase quantitation.