Learn More
Biotium One-Step Lumitein™ UV Protein Gel Stain, 1X
Catalog No. 501964386
Encompass_Preferred
One-Step Lumitein™ UV Protein Gel Stain is a ready-to-use luminescent protein gel stain. Gels stained with One-Step Lumitein™ UV can be visualized with a variety of different UV-based fluorescence imaging systems. Convenient staining protocol, requiring only a single 5-30 minute staining step without fixation, destaining is optional. The stain is sensitive, detecting ~ 1-10 ng of protein per band (staining intensity can vary between proteins) and comparable to the Oriole™ Fluorescent Gel Stain(Bio-Rad). Unlike other fluorescent gel stains like Oriole™ or Flamingo™, One-Step Lumitein™ UV is an aqueous-based solution that does not does not contain hazardous methanol or acetic acid for safer handling and ease of disposal, does not cause gel shrinkage, and is significantly lower in cost. Staining is fully compatible with mass spectrometry and Edman-based sequencing.
Catalog No. 50-196-4386 Supplier Biotium Supplier No. 210054L
One-Step Lumitein™ UV Protein Gel Stain is a ready-to-use luminescent protein gel stain. Gels stained with One-Step Lumitein™ UV can be visualized with a variety of different UV-based fluorescence imaging systems. Convenient staining protocol, requiring only a single 5-30 minute staining step without fixation, destaining is optional. The stain is sensitive, detecting ~ 1-10 ng of protein per band (staining intensity can vary between proteins) and comparable to the Oriole™ Fluorescent Gel Stain(Bio-Rad). Unlike other fluorescent gel stains like Oriole™ or Flamingo™, One-Step Lumitein™ UV is an aqueous-based solution that does not does not contain hazardous methanol or acetic acid for safer handling and ease of disposal, does not cause gel shrinkage, and is significantly lower in cost. Staining is fully compatible with mass spectrometry and Edman-based sequencing.