Learn More
Roche Diagnostics KAPA SYBR FAST One-Step qRT-PCR Kits - 500 x 20 uL reactions (Universal), 5 mL
Catalog No. 501965200 Shop All Roche Diagnostics Products
Encompass
Kits include KAPA RT Mix, dUTP, and KAPA SYBR FAST qPCR Master Mix, which contains KAPA SYBR FAST DNA Polymerase, reaction buffer, dNTPs, SYBR Green I dye, and MgCl2 at a final concentration of 2.5 mM. *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5200 Supplier Roche Diagnostics Supplier No. 07959621001
Kits include KAPA RT Mix, dUTP, and KAPA SYBR FAST qPCR Master Mix, which contains KAPA SYBR FAST DNA Polymerase, reaction buffer, dNTPs, SYBR Green I dye, and MgCl2 at a final concentration of 2.5 mM. *This item is non returnable.