Learn More
INTACT GENOMICS INC igScript™ One Step RT-PCR Kit 500 Reactions
Catalog No. 5000083027
Encompass
igScript™ One Step RT-PCR kit combines two powerful mixtures: i) igScript™ Reverse Transcriptase and ii) Hot start Taq 2x master mix providing improved PCR efficiency, wider dynamic range, superior sensitivity and specificity. The two mixtures require minimal handling during reaction setup and offer consistent and robust RT-PCR reactions.Applications: Gene expression data validation. Multiplexing Mutation detection Pathogen and viral detection Genetically modified organisms (GMO) characterization and Genetic profiling Benefits: Enhanced efficiency, specificity, and sensitivity Compatible with all real-time PCR instruments Superior gene expression results under various cycling conditions Robust and active for cDNA synthesis at temperatures up to 55°C.
Catalog No. 50-000-83027 Supplier INTACT GENOMICS INC Supplier No. 4213
igScript™ One Step RT-PCR kit combines two powerful mixtures: i) igScript™ Reverse Transcriptase and ii) Hot start Taq 2x master mix providing improved PCR efficiency, wider dynamic range, superior sensitivity and specificity. The two mixtures require minimal handling during reaction setup and offer consistent and robust RT-PCR reactions.Applications: Gene expression data validation. Multiplexing Mutation detection Pathogen and viral detection Genetically modified organisms (GMO) characterization and Genetic profiling Benefits: Enhanced efficiency, specificity, and sensitivity Compatible with all real-time PCR instruments Superior gene expression results under various cycling conditions Robust and active for cDNA synthesis at temperatures up to 55°C.