Learn More
VAZYME INTERNATIONAL LLC Vazyme International AccurSTART II U+ One Step RT-qPCR Super Premix, Fast, One Tube
Catalog No. NC3979695
Encompass
This preassembled, probe-based master mix is engineered for one-tube, one-step RT-qPCR applications. It is optimized for rapid and sensitive quantification of RNA templates in both singleplex and multiplex assays, such as for RNA viruses.
  • Fully preassembled and stable, compatible with preloaded primers and probes, ideal for long-term storage.
  • Ultra-fast amplification with reliable detection within 22 minutes, maintaining sensitivity.
  • Direct amplification compatibility, supporting high-volume input of crude lysates without purification.
  • High-sensitivity multiplexing for accurate detection of low-copy viral RNA across multiple targets.
  • Engineered hot-start enzyme system ensures precise activation and minimizes non-specific amplification.
  • dUTP/UDG contamination control system functions at room temperature, preventing carryover.
Catalog No. NC3979695 Supplier VAZYME INTERNATIONAL LLC Supplier No. Q63101
This preassembled, probe-based master mix is engineered for one-tube, one-step RT-qPCR applications. It is optimized for rapid and sensitive quantification of RNA templates in both singleplex and multiplex assays, such as for RNA viruses.
  • Fully preassembled and stable, compatible with preloaded primers and probes, ideal for long-term storage.
  • Ultra-fast amplification with reliable detection within 22 minutes, maintaining sensitivity.
  • Direct amplification compatibility, supporting high-volume input of crude lysates without purification.
  • High-sensitivity multiplexing for accurate detection of low-copy viral RNA across multiple targets.
  • Engineered hot-start enzyme system ensures precise activation and minimizes non-specific amplification.
  • dUTP/UDG contamination control system functions at room temperature, preventing carryover.