Learn More
Zymo Research Corporation One-Way Luer-Lock Clear Stopcock, 20 pack
SDP
Catalog No. NC1079010
Encompass_Preferred
One-way Luer-Lock Stopcock to be used with the EZ-Vac™ Vacuum Manifold. For use with Zymo Research purification columns.
Catalog No. NC1079010 Supplier Zymo Research Corporation Supplier No. S7001
One-way Luer-Lock Stopcock to be used with the EZ-Vac™ Vacuum Manifold. For use with Zymo Research purification columns.