Learn More
Betty Mills Company ONE WAY STOP COCKS 20CASE
SDP
Catalog No. NC2385724
Encompass
Externally Operated valve helps regulate the flow of liquid or gas.Can be attached to any configuration inside a pack for custom procedures.Responsive on/off valve is designed to maximize20 per caseHandle-offLuer Type-Luer SlipNon-Sterile Component-SPT-STOP1SLIPPressure Rating-45 PSISterile-YesStopcock Type- 1-WAY
Catalog No. NC2385724 Supplier Betty Mills Company Supplier No. MED DYNJSC1MPFLS
Externally Operated valve helps regulate the flow of liquid or gas.Can be attached to any configuration inside a pack for custom procedures.Responsive on/off valve is designed to maximize20 per caseHandle-offLuer Type-Luer SlipNon-Sterile Component-SPT-STOP1SLIPPressure Rating-45 PSISterile-YesStopcock Type- 1-WAY