Learn More
Aviva Systems Biology Corp ONECUT1 Antibody - middle region (ARP38481_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38481_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ONECUT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ONECUT1. Alternate names: HNF-6, HNF6, HNF6AApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53106