Learn More
Aviva Systems Biology Corp ONECUT1 Antibody - N-terminal region (ARP38480_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38480_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ONECUT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ONECUT1. Alternate names: HNF-6, HNF6, HNF6AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56670