Learn More
Aviva Systems Biology Corp ONECUT2 Antibody - middle region (ARP57926_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57926_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ONECUT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ONECUT2. Alternate names: MGC120377, MGC120378, OC-2, OC2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62180