Learn More
Aviva Systems Biology Corp Onecut3 Antibody - C-terminal region (ARP37481_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37481_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Onecut3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the c terminal region of mouse Onecut3. Alternate names: OC-3, Oc3ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-641