Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ OneFeed™ Supplement
GREENER_CHOICE

Catalog No. 670108
Change view
Click to view available options
Quantity:
10 L
50 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-010-8 50 L
67-010-9 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 67-010-8 Supplier Gibco™ Supplier No. 670108
Only null left
Add to Cart
Add to Cart

A versatile medium supplement.

Gibco™ OneFeed™ Supplement is a chemically-defined, animal-free, protein-free feed shown to enhance protein production when used in a variety of commercially available medium.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Cell Line CHO DG44, BHK-21, CHO-K1, CHO
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder
Product Type Media Supplement
Format BPC
Quantity 50 L
Source Animal origin-free
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.