Learn More
Alkali Scientific OneFit 10uL Extra Long Univers
SDP
Catalog No. 503026917
Encompass_Preferred
OneFitmultichannel filter tips are the ultimate solution for high-throughput screening (HTS) applications These tips feature an innovative soft and stretchable TPE resin collar that creates a flawless seal guaranteeing unparalleled performance and accuracy with each use The reduced insertion forces make sealing and ejection effortless on multichannel pipettes streamlining your workflow Designed to fit any pipettor OneFitensures ergonomic comfort during use enhancing efficiency in the lab Additionally the soft seal and low insertion force significantly reduce pipette cone wear and erosion extending the lifespan of your multi-channel pipette While the collar is soft and flexible the rest of the tip is made from rigid resin ensuring crystal-clear straight tips for precise dispensing For added convenience and confidence OneFitmultichannel filter tips are graduated for volume confirmation and come in extended volume sizes ideal for reducing contamination risks in sensitive assays Cer
Catalog No. 50-302-6917 Supplier Alkali Scientific Supplier No. FT9010CS
OneFitmultichannel filter tips are the ultimate solution for high-throughput screening (HTS) applications These tips feature an innovative soft and stretchable TPE resin collar that creates a flawless seal guaranteeing unparalleled performance and accuracy with each use The reduced insertion forces make sealing and ejection effortless on multichannel pipettes streamlining your workflow Designed to fit any pipettor OneFitensures ergonomic comfort during use enhancing efficiency in the lab Additionally the soft seal and low insertion force significantly reduce pipette cone wear and erosion extending the lifespan of your multi-channel pipette While the collar is soft and flexible the rest of the tip is made from rigid resin ensuring crystal-clear straight tips for precise dispensing For added convenience and confidence OneFitmultichannel filter tips are graduated for volume confirmation and come in extended volume sizes ideal for reducing contamination risks in sensitive assays Cer