Learn More
Alkali Scientific OneFit Universal Soft Touch Fi
SDP
Supplier:  Alkali Scientific ST91000
Encompass_Preferred
OneFitmultichannel friendly pipette tips stand out as the ideal choice for HTS work Featuring an innovative soft and stretchable TPE resin collar these tips create a flawless seal ensuring unmatched performance and accuracy every time they are used Insertion forces are significantly reduced making sealing and ejection effortless on multichannel pipettes OneFits design seamlessly transforms any pipettor into an ergonomic tool enhancing comfort during use Moreover the soft seal and low insertion force contribute to a notable reduction in pipette cone wear and erosion thereby prolonging the lifespan of your multi-channel pipette While the collar boasts a soft material the remainder of the pipette tip is crafted from rigid resin resulting in crystal-clear straight tips that facilitate precise dispensing For added convenience OneFitmultichannel tips are graduated for volume confirmation and come in extended volume sizes which are preferred for minimizing contamination risks Furthermo
Catalog No. 50-302-8198