Learn More
Perkin Elmer US LLC One-Piece Quartz Torch, 1.0 mm I.D. Injector - Triple Blue Mark
Catalog No. 502113280
Encompass_Preferred
This one-piece quartz torch, with three blue marks, has a 1.0 mm I.D. injector integrated directly into the torch. It also has a PVDF o-ring free holder. The torch is an economical solution for many non-HF applications and ideal for organic solvent analysis. The one-piece Torch-Injector design allows for easy maintenance and quick torch alignment.
Catalog No. 50-211-3280 Supplier Perkin Elmer US LLC Supplier No. N8152699
This one-piece quartz torch, with three blue marks, has a 1.0 mm I.D. injector integrated directly into the torch. It also has a PVDF o-ring free holder. The torch is an economical solution for many non-HF applications and ideal for organic solvent analysis. The one-piece Torch-Injector design allows for easy maintenance and quick torch alignment.