Learn More
Perkin Elmer US LLC One-Piece Quartz Torch, 1.5 mm I.D. Injector - Double Blue Mark
Catalog No. 502111903
Encompass_Preferred
This one-piece quartz torch, marked double blue, with a PVDF o-ring free holder, has a 1.5 mm I.D. injector integrated directly into the torch. This design allows for easy maintenance and quick torch alignment.
Catalog No. 50-211-1903 Supplier Perkin Elmer US LLC Supplier No. N8152427
This one-piece quartz torch, marked double blue, with a PVDF o-ring free holder, has a 1.5 mm I.D. injector integrated directly into the torch. This design allows for easy maintenance and quick torch alignment.