Learn More
Perkin Elmer US LLC One-Piece Quartz Torch, 2.0 mm I.D. Injector - Blue/Purple Mark
Catalog No. 502103868
Encompass_Preferred
This one-piece quartz torch, with a PVDF o-ring free holder, has a 2.0 mm injector integrated directly into the torch. This design allows for easy maintenance and quick torch alignment. Included in: NexION 2000 ICP-MS Blue Base Configuration's Sample Introduction Kit (N8150034), Purple NexION 1000 ICP-MS Sample Introduction Kit (N8150038), NexION 2000 B Consumables Kit (N8150054), and NexION 1000 Consumables Kit (N8150058).
Catalog No. 50-210-3868 Supplier Perkin Elmer US LLC Supplier No. N8152472
This one-piece quartz torch, with a PVDF o-ring free holder, has a 2.0 mm injector integrated directly into the torch. This design allows for easy maintenance and quick torch alignment. Included in: NexION 2000 ICP-MS Blue Base Configuration's Sample Introduction Kit (N8150034), Purple NexION 1000 ICP-MS Sample Introduction Kit (N8150038), NexION 2000 B Consumables Kit (N8150054), and NexION 1000 Consumables Kit (N8150058).