Learn More
Perkin Elmer US LLC One-Piece Quartz Torch, 2.5 mm I.D. Injector - Green Mark
Catalog No. 502103865
Encompass_Preferred
This one-piece quartz torch, marked green, with a PVDF o-ring free holder, has a 2.0 mm injector integrated directly into the torch. This design allows for easy maintenance and quick torch alignment. Included in: NexION 2000 ICP-MS Core Configuration's Sample Introduction Kit (N8150034) and NexION 2000 C Consumables Kit (N8150055) .
Catalog No. 50-210-3865 Supplier Perkin Elmer US LLC Supplier No. N8152473
This one-piece quartz torch, marked green, with a PVDF o-ring free holder, has a 2.0 mm injector integrated directly into the torch. This design allows for easy maintenance and quick torch alignment. Included in: NexION 2000 ICP-MS Core Configuration's Sample Introduction Kit (N8150034) and NexION 2000 C Consumables Kit (N8150055) .