Learn More
Perkin Elmer US LLC One-Piece SilQ Torch, 2.0 mm I.D. Injector - White Mark
Catalog No. 502111893
Encompass_Preferred
This one-piece SilQ torch, marked white, with a PVDF o-ring free holder, has a 2.0 mm injector integrated directly into the torch. This design allows for easy maintenance and quick torch alignment. Included in: the Semi-conductor Configuration's Sample Intruduction Kit (N8150037), the NexION 1000/2000 Consumables Kit for NexION S Configuration (N8150057) and the Single Cell Sample Introduction Kit for NexION 1000/2000 (N8150032).
Catalog No. 50-211-1893 Supplier Perkin Elmer US LLC Supplier No. N8152428
This one-piece SilQ torch, marked white, with a PVDF o-ring free holder, has a 2.0 mm injector integrated directly into the torch. This design allows for easy maintenance and quick torch alignment. Included in: the Semi-conductor Configuration's Sample Intruduction Kit (N8150037), the NexION 1000/2000 Consumables Kit for NexION S Configuration (N8150057) and the Single Cell Sample Introduction Kit for NexION 1000/2000 (N8150032).