Learn More
Research Products International Corp OneRack Multi, Purple
SDP
Supplier:  Research Products International Corp 248546
Encompass
The most flexible tube rack designed for use with a multitude of tube diameters and heights. Multiple configurations allow interchangeable tube rack inserts to hold multiple tube sizes firmly in place. Inserts can be pivoted in either an upper or lower position to securely accommodate the size and height of tube.Unique multi-functional design allows tube rack insert to rotate to an angle position to facilitate more comfortable pipetting and allow better visibility. Rack wells have a unique teardrop design that enables better retention of tubes over traditional designs. Ergonimic handles transport rack easily and alphanumeric grid quickly identifies sample location. Manufactured of autoclavable polypropylene.
Mutli-functional design
Interchangeable tube rack inserts
Fits 10-30mm tubes
Rotate tubes for accessibility
Catalog No. 50-225-2998