Learn More
Research Products International Corp OneStep-96 PCR Inhibitor Removal Kit, 2 x 96 Preps
SDP
Catalog No. 501977247
Encompass
The OneStep PCR Inhibitor Removal Kit contains all the components needed for efficient removal of contaminants that can inhibit downstream enzymatic reactions (e.g. PCR and RT) from DNA and RNA preparations. The column matrices have been specifically designed for the efficient removal of polyphenolic compounds, humic/fulvic acids, tannins, melanin, etc. from the most impure DNA and RNA preparations. Sample clean-up is as simple as applying, spinning, and recovering a sample from the column.
Removes PCR inhibitors such as polyphenolics, humic/fulvic acids, tannins, melanin, etc. from nucleic acid solutions to yield high quality DNA or RNA.
Fast, one-step procedure for cleaning impure samples prior to PCR, sequencing, reverse transcription (RT), etc.
Catalog No. 50-197-7247 Supplier Research Products International Corp Supplier No. ZD6035
The OneStep PCR Inhibitor Removal Kit contains all the components needed for efficient removal of contaminants that can inhibit downstream enzymatic reactions (e.g. PCR and RT) from DNA and RNA preparations. The column matrices have been specifically designed for the efficient removal of polyphenolic compounds, humic/fulvic acids, tannins, melanin, etc. from the most impure DNA and RNA preparations. Sample clean-up is as simple as applying, spinning, and recovering a sample from the column.
Removes PCR inhibitors such as polyphenolics, humic/fulvic acids, tannins, melanin, etc. from nucleic acid solutions to yield high quality DNA or RNA.
Fast, one-step procedure for cleaning impure samples prior to PCR, sequencing, reverse transcription (RT), etc.