Learn More
Zymo Research Corporation OneStep PCR Inhibitor Removal
SDP
Catalog No. 502865904
Encompass_Preferred
TheOneStepPCR Inhibitor Removal kits are PCR inhibitor clean up kits that contain all the components needed for efficient removal of contaminants from DNA and RNA preparations that can inhibit downstream enzymatic reactions (e.g. PCR and RT).The column slurry has been optimized to efficiently bind and remove any polyphenolic compounds, such as humic/fulvic acids, tannins, melanin, etc. from the most impure DNA and RNA preparations.Sampleclean-upis as simple as filtering your sample with the prepped column.
Catalog No. 50-286-5904 Supplier Zymo Research Corporation Supplier No. D6031
TheOneStepPCR Inhibitor Removal kits are PCR inhibitor clean up kits that contain all the components needed for efficient removal of contaminants from DNA and RNA preparations that can inhibit downstream enzymatic reactions (e.g. PCR and RT).The column slurry has been optimized to efficiently bind and remove any polyphenolic compounds, such as humic/fulvic acids, tannins, melanin, etc. from the most impure DNA and RNA preparations.Sampleclean-upis as simple as filtering your sample with the prepped column.