Learn More
Zymo Research Corporation OneStep™ PCR Inhibitor Removal Kit (50 Preps)
SDP
Catalog No. 50444470
Encompass_Preferred
The column matrix has been specifically designed for efficient removal of polyphenolic compounds, humic/fulvic acids, tannins, melanin, etc. from most impure DNA and RNA preparations. Sample cleanup is as simple as: applying, spinning and recovering asample from the column.
Catalog No. 50-444-470 Supplier Zymo Research Corporation Supplier No. D6030
The column matrix has been specifically designed for efficient removal of polyphenolic compounds, humic/fulvic acids, tannins, melanin, etc. from most impure DNA and RNA preparations. Sample cleanup is as simple as: applying, spinning and recovering asample from the column.