Learn More
Zymo Research Corporation OneStep PLUS qMethyl PCR Kit
SDP
Catalog No. 502643407
Encompass_Preferred
PCR Kit No bisulfite conversion needed from DNA sample to results in just one step. Accurate quantification of DNA methylation levels at the genomic regions of choice. Ideal for rapid screening of DNA methylation at single and multi-locus. 1/EA
Catalog No. 50-264-3407 Supplier Zymo Research Corporation Supplier No. D5312
PCR Kit No bisulfite conversion needed from DNA sample to results in just one step. Accurate quantification of DNA methylation levels at the genomic regions of choice. Ideal for rapid screening of DNA methylation at single and multi-locus. 1/EA