Learn More
Novus Biologicals™ OneStep TUNEL Apoptosis Kit, Red, 555

Specifications
Specifications
| Description | OneStep TUNEL |
| For Use With (Application) | Flow Cytometry, Fluorescence Imaging |
| Content And Storage | Store at -20C. |
| Quantity | 20 Assays |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.